Novus Biologicals
Manufacturer Code:NBP275965
Catalog # NBP275965
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-YLRRFFKAKKLIE corresponding to C-Terminus according to NP_006818.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
Protein Aliases: 21 kDa transmembrane trafficking protein; 21 kDa transmembrane-trafficking protein; 21 kDa transmembrane-trafficking protein p23 P24(DELTA) p24delta S31I125 S31III125 TMP21Tmp-21-I transmembrane emp24 domain-containing protein 10 transmembrane emp24-like trafficking protein 10 (yeast) Transmembrane protein Tmp2121 kDa transmembrane trafficking protein; p23; p24 family protein delta-1; p24delta; p24delta1; Protein TMED10; S31I125; S31III125; testicular tissue protein Li 206; Tmp-21-I; Transmembrane emp24 domain-containing protein 10; transmembrane emp24-like trafficking protein 10; Transmembrane protein Tmp21
Gene Aliases: p23; P24(DELTA); p24d1; S31I125; S31III125; TMED10; Tmp-21-I; TMP21
UniProt ID: (Human) P49755
Entrez Gene ID: (Human) 10972
Molecular Function: membrane traffic protein transfer/carrier protein vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.