Invitrogen
Manufacturer Code:PA568473
Catalog # PIPA568473
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human Titin &lsqbaa 161-210]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMD1G CMH9 CMPD4 CONNECTIN DKFZp451N061 EOMFC FLJ26020 FLJ26409 FLJ32040 FLJ34413 FLJ39564 FLJ43066 HMERF LGMD2J TMD; Connectin; Rhabdomyosarcoma antigen MU-RMS-40.14; Titin; titin protein homolog; ttn
Gene Aliases: 1100001C23Rik; 2310036G12Rik; 2310057K23Rik; 2310074I15Rik; AF006999; AV006427; CMD1G; CMH9; CMPD4; D330041I19Rik; D830007G01Rik; EOMFC; HMERF; L56; LGMD2J; mdm; MYLK5; shru; TMD; TTN
UniProt ID: (Human) Q8WZ42
Entrez Gene ID: (Human) 7273, (Rat) 84015, (Mouse) 22138
Molecular Function:
cell adhesion molecule
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.