Novus Biologicals
Manufacturer Code:NBP188071
Catalog # NBP188071
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GTVSTSCYLAVQVSEEFEKETTAVTEKFTTEEKRFVESRDVVMTDTSLTEEQAGPGEPAAPYFITKPVVQKLVEGGSVVFGCQVGGNPKPHVYWKKSGVPLTTGYRYKVSYNKQTGECKLVISMTFADDAGEYT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cardiomyopathy dilated 1G (autosomal dominant) CMD1G CMH9 CMPD4 Connectin DKFZp451N061 EC 2.7.11.1 EOMFC FLJ32040 FLJ34413 FLJ39564 FLJ43066 HMERF LGMD2JFLJ26020 MPRM Rhabdomyosarcoma antigen MU-RMS-40.14 titin TMDFLJ26409; Connectin; Rhabdomyosarcoma antigen MU-RMS-40.14; Titin
Gene Aliases: CMD1G; CMH9; CMPD4; EOMFC; HMERF; LGMD2J; MYLK5; TMD; TTN
UniProt ID: (Human) Q8WZ42
Entrez Gene ID: (Human) 7273
Molecular Function:
cell adhesion molecule
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.