Novus Biologicals
Manufacturer Code:NB10040853
Catalog # NB10040853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1170 and the C-terminus (residue 1208) of human Timeless using the numbering given in entry NP_003911.1 (GeneID 8914). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ12640 hTIMTIMELESS1 TIM1protein timeless homolog timeless (Drosophila) homolog timeless homolog (Drosophila) TIMFLJ20714; hTIM; Protein timeless homolog; timeless circadian clock 1; timeless homolog; Tof1 homolog
Gene Aliases: hTIM; TIM; TIM1; TIMELESS; TIMELESS1
UniProt ID: (Human) Q9UNS1
Entrez Gene ID: (Human) 8914
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.