Proteintech
Manufacturer Code:111231AP150UL
Catalog # 50-172-6895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Tim23 Fusion Protein Ag1620 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mitochondrial import inner membrane translocase subunit Tim23; PRO1197 TIM23 TIMM23; translocase of inner mitochondrial membrane 23 homolog; translocase of the inner mitochondrial membrane
Gene Aliases: TIM23; TIMM23
UniProt ID: (Human) O14925, (Rat) O35093
Entrez Gene ID: (Human) 100287932, (Rat) 54312, (Mouse) 53600
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.