Novus Biologicals
Manufacturer Code:NBP220641
Catalog # NBP220641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human TJP2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C9DUPq21.11 DFNA51 DUP9q21.11 MGC26306 Tight junction protein 2 tight junction protein 2 (zona occludens 2) TJP2 X104 X104tight junction protein ZO-2 ZO2 ZO-2 ZO2Friedreich ataxia region gene X104 (tight junction protein ZO-2) Zona occludens protein 2 Zonula occludens protein 2; Friedreich ataxia region gene X104 (tight junction protein ZO-2); Tight junction protein 2; Tight junction protein ZO-2; zona occludens 2; Zona occludens protein 2; Zonula occludens protein 2
Gene Aliases: C9DUPq21.11; DFNA51; DUP9q21.11; PFIC4; TJP2; X104; ZO2
UniProt ID: (Human) Q9UDY2
Entrez Gene ID: (Human) 9414
Molecular Function:
cell junction protein
tight junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.