Novus Biologicals
Manufacturer Code:NBP298900AF647
Catalog # NB056391
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Tight Junction Protein 2. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C9DUPq21.11 DFNA51 DUP9q21.11 MGC26306 Tight junction protein 2 tight junction protein 2 (zona occludens 2) TJP2 X104 X104tight junction protein ZO-2 ZO2 ZO-2 ZO2Friedreich ataxia region gene X104 (tight junction protein ZO-2) Zona occludens protein 2 Zonula occludens protein 2; Friedreich ataxia region gene X104 (tight junction protein ZO-2); Tight junction protein 2; Tight junction protein ZO-2; zona occludens 2; Zona occludens protein 2; Zonula occludens protein 2
Gene Aliases: C9DUPq21.11; DFNA51; DUP9q21.11; PFIC4; TJP2; X104; ZO2
UniProt ID: (Human) Q9UDY2
Entrez Gene ID: (Human) 9414
Molecular Function: cell junction protein tight junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.