R&D Systems
Manufacturer Code:AF313SP
Catalog # AF313SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Tie-2 Ala23-Lys745 Accession # Q02763 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Angiopoietin-1 receptor; angiopoietin-1 receptor CD202B CD202b antigen EC 2.7.10 EC 2.7.10.1 hTIE2 p140 TEK soluble TIE2 variant 1 soluble TIE2 variant 2 TEK TEK tyrosine kinase endothelial Tie2 TIE-2 TIE2CD202b Tunica interna endothelial cell kinase Tyrosine-protein kinase receptor TEK Tyrosine-protein kinase receptor TIE-2 venous malformations multiple cutaneous and mucosal VMCM VMCM1; CD202b; Endothelial tyrosine kinase; hTIE2; p140 TEK; TEK tyrosine kinase, endothelial; Tunica interna endothelial cell kinase; Tyrosine kinase with Ig and EGF homology domains-2; Tyrosine-protein kinase receptor TEK; Tyrosine-protein kinase receptor TIE-2
Gene Aliases: CD202B; TEK; TIE-2; TIE2; VMCM; VMCM1
UniProt ID: (Human) Q02763
Entrez Gene ID: (Human) 7010
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.