Novus Biologicals
Manufacturer Code:NBP237267
Catalog # NBP237267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8D12D2 |
Immunogen | Purified recombinant fragment of human TIE1 (AA: 385-607) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.1 JTK14soluble TIE1 variant 3 receptor tyrosine kinase soluble TIE1 variant 1 soluble TIE1 variant 2 soluble TIE1 variant 5 TIEsoluble TIE1 variant 4 tyrosine kinase with immunoglobulin and epidermal growth factor homologydomains 1 tyrosine kinase with immunoglobulin-like and EGF-like domains 1 tyrosine-protein kinase receptor Tie-1; tyrosine kinase with immunoglobulin and epidermal growth factor homology domains 1; tyrosine kinase with immunoglobulin-like and EGF-like domains 1; Tyrosine-protein kinase receptor Tie-1
Gene Aliases: JTK14; TIE; TIE1
UniProt ID: (Human) P35590
Entrez Gene ID: (Human) 7075
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.