Novus Biologicals
Manufacturer Code:NBP295244100UL
Catalog # NB024650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 25-230 of human TRH (NP_009048.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC125964 MGC125965 prothyroliberin thyrotropin-releasing hormone; Pro-thyrotropin-releasing hormone; Pro-TRH; Prothyroliberin; Protirelin; Thyroliberin; Thyrotropin-releasing factor; Thyrotropin-releasing hormone; TRF; TRH; TSH-releasing factor
Gene Aliases: Pro-TRH; TRF; TRH
UniProt ID: (Human) P20396
Entrez Gene ID: (Human) 7200
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.