Novus Biologicals
Manufacturer Code:NBP233450
Catalog # NBP233450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: LNSGNVRHRLQFYIGEHLLPYNMTVYQAVRQFSIQAEDERESTDDESNPLGRAGIWTKTHTIWYKPVREDEESNKDCVGGKRGRAQTAPTKTSP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase for Arf; E3 ubiquitin-protein ligase TRIP12; EC 6.3.2.- KIAA0045ULF MGC138849 MGC138850 probable E3 ubiquitin-protein ligase TRIP12 thyroid hormone receptor interactor 12 thyroid receptor interacting protein 12 Thyroid receptor-interacting protein 12 TR-interacting protein 12 TRIP-12; HECT-type E3 ubiquitin transferase TRIP12; probable E3 ubiquitin-protein ligase TRIP12; thyroid receptor interacting protein 12; Thyroid receptor-interacting protein 12; TR-interacting protein 12; ULF
Gene Aliases: KIAA0045; TRIP-12; TRIP12; ULF
UniProt ID: (Human) Q14669
Entrez Gene ID: (Human) 9320
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.