R&D Systems
Manufacturer Code:MAB819SP
Catalog # MAB819SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 60000000000 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Thymopoietin aa 2-187 Accession # P42166 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CMD1T lamina-associated polypeptide 2 LAP2PRO0868 LEM domain containing 4 LEMD4 MGC61508 thymopoietin Thymopoietin isoform alpha Thymopoietin isoforms beta/gamma Thymopoietin-related peptide isoform alpha Thymopoietin-related peptide isoforms beta/gamma TMPO TP TP alpha TP beta/gamma TPRP isoform alpha TPRP isoforms beta/gamma; lamina-associated polypeptide 2; Lamina-associated polypeptide 2, isoform alpha; Lamina-associated polypeptide 2, isoforms beta/gamma; LEM domain containing 4; Splenin; Thymopentin; Thymopoietin; thymopoietin beta; Thymopoietin isoform alpha; Thymopoietin, isoforms beta/gamma; Thymopoietin-related peptide isoform alpha; Thymopoietin-related peptide isoforms beta/gamma; TP; TP alpha; TP beta/gamma; TP5; TPRP isoform alpha; TPRP isoforms beta/gamma
Gene Aliases: CMD1T; LAP2; LEMD4; PRO0868; TMPO; TP
UniProt ID: (Human) P42166
Entrez Gene ID: (Human) 7112
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.