Novus Biologicals
Manufacturer Code:NBP234502AF488
Catalog # NBP234502X
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TS106 |
Immunogen | Recombinant human thymidylate synthase |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1.45 HsT422 MGC88736 thymidylate synthase thymidylate synthetase TMSTsase TSase TSHST422; Thymidylate synthase; TS; TSase
Gene Aliases: HST422; OK/SW-cl.29; TMS; TS; TYMS
UniProt ID: (Human) P04818
Entrez Gene ID: (Human) 7298
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.