Novus Biologicals
Manufacturer Code:NBP17989020UL
Catalog # NBP17989020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The specific Immunogen is proprietary information. Peptide sequence HLHEEWLIKGSLFPMAAPVLVIEADHHMERMLELFEQNRDRILTPENRKH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.1.21 MTDPS2 Mt-TK MTTK thymidine kinase 2 mitochondrial; Mt-TK; Thymidine kinase 2, mitochondrial
Gene Aliases: MTDPS2; MTTK; SCA31; TK2
UniProt ID: (Human) O00142
Entrez Gene ID: (Human) 7084
Molecular Function: kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.