R&D Systems
Manufacturer Code:MAB8180100
Catalog # MAB8180100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1008515 |
Immunogen | <em>E. coli</em>-derived recombinant human Thymidine Kinase 1 Met1-Asn234 Accession # P04183 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.7.1.21 thymidine kinase 1 soluble isoform thymidine kinase 1 soluble thymidine kinase cytosolic thymidine kinase-1 TK1 TK2; thymidine kinase 1 soluble isoform; thymidine kinase 1, soluble; Thymidine kinase, cytosolic; thymidine kinase-1
Gene Aliases: TK1; TK2
UniProt ID: (Human) P04183
Entrez Gene ID: (Human) 7083
Molecular Function:
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.