Invitrogen
Manufacturer Code:MA525163
Catalog # PIMA525163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A9 |
Immunogen | Full length human recombit protein of TBXAS1 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BDPLT14 CYP5 CYP5A1 Cytochrome P450 5A1 Cytochrome P450 family 5 subfamily A polypeptide 1 FLJ52771 GHOSAL Platelet cytochrome P450 subfamily V TBXAS1 THAS THAS&lowbarHUMAN Thromboxane A synthase 1 (platelet) Thromboxane A synthase 1 (platelet cytochrome P450 family 5 subfamily A) Thromboxane A synthase 1 Thromboxane A synthase 1 platelet cytochrome P450 subfamily V Thromboxane-A synthase TS TXA synthase TXAS TXS; Cytochrome P450 5A1; cytochrome P450, family 5, subfamily A, polypeptide 1; Hydroperoxy icosatetraenoate dehydratase; platelet, cytochrome P450, subfamily V; thromboxane A synthase 1 (platelet); Thromboxane-A synthase; TXA synthase
Gene Aliases: BDPLT14; CYP5; CYP5A1; GHOSAL; TBXAS1; THAS; TS; TXAS; TXS
UniProt ID: (Human) P24557
Entrez Gene ID: (Human) 6916
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.