Invitrogen
Manufacturer Code:PA585175
Catalog # PIPA585175
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human TXA synthase. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYP5 antibody CYP5A1 antibody FLJ52771 antibody GHOSAL antibody THAS antibody TS antibody TXAS antibody TXS antibody TBXAS1 antibody platelet cytochrome P450 subfamily V antibody TXA synthase antibody thromboxane-A synthase antibody thromboxane A synthase 1 (platelet cytochrome P450 family 5 subfamily A) antibody cytochrome P450 5A1 antibody cytochrome P450 family 5 subfamily A polypeptide 1 antibody thromboxane A synthase 1 (platelet) antibody; Cytochrome P450 5A1; Hydroperoxy icosatetraenoate dehydratase; ThromboxA ane synthase 1; Thromboxane-A synthase; TXA synthase; TXS
Gene Aliases: Cyp5; Cyp5a1; Tbxas1
UniProt ID: (Rat) P49430
Entrez Gene ID: (Rat) 24886
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.