Invitrogen
Manufacturer Code:PA576625
Catalog # PIPA576625
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human TBXAS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BDPLT14 CYP5 CYP5A1 Cytochrome P450 5A1 Cytochrome P450 family 5 subfamily A polypeptide 1 FLJ52771 GHOSAL Platelet cytochrome P450 subfamily V TBXAS1 THAS THAS&lowbarHUMAN Thromboxane A synthase 1 (platelet) Thromboxane A synthase 1 (platelet cytochrome P450 family 5 subfamily A) Thromboxane A synthase 1 Thromboxane A synthase 1 platelet cytochrome P450 subfamily V Thromboxane-A synthase TS TXA synthase TXAS TXS; Cytochrome P450 5A1; cytochrome P450, family 5, subfamily A, polypeptide 1; Hydroperoxy icosatetraenoate dehydratase; platelet, cytochrome P450, subfamily V; thromboxane A synthase 1 (platelet); Thromboxane-A synthase; TS; TXA synthase
Gene Aliases: BDPLT14; CYP5; CYP5A1; GHOSAL; TBXAS1; THAS; TS; TXAS; TXS
UniProt ID: (Human) P24557, (Mouse) P36423
Entrez Gene ID: (Human) 6916, (Mouse) 21391
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.