Novus Biologicals
Manufacturer Code:NBP233946
Catalog # NBP233946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: RGALMSAFSPEKLNEMVPLISQACDLLLAHLKRYAESGDAFDIQRCYCNYTTDVVASVAFGTPVDSWQAPEDPFVKHCKRFFEFCIP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYP5A1family 5 subfamily A polypeptide 1 CYP5FLJ52771 EC 5.3.99.5 GHOSAL platelet cytochrome P450 subfamily V thromboxane A synthase 1 (platelet) thromboxane A synthase 1 (platelet cytochrome P450 family 5 subfamily A) thromboxane A synthase 1 (platelet cytochrome P450 subfamily V) thromboxane-A synthase TXA synthase; Cytochrome P450 5A1; cytochrome P450, family 5, subfamily A, polypeptide 1; Hydroperoxy icosatetraenoate dehydratase; platelet, cytochrome P450, subfamily V; thromboxane A synthase 1 (platelet); Thromboxane-A synthase; TXA synthase
Gene Aliases: BDPLT14; CYP5; CYP5A1; GHOSAL; TBXAS1; THAS; TS; TXAS; TXS
UniProt ID: (Human) P24557
Entrez Gene ID: (Human) 6916
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.