Invitrogen
Manufacturer Code:PA545676
Catalog # PIPA545676
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human TBXAS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytochrome P450 5A1; cytochrome P450, family 5, subfamily A, polypeptide 1; Hydroperoxy icosatetraenoate dehydratase; platelet, cytochrome P450, subfamily V; ThromboxA ane synthase 1; thromboxane A synthase 1 (platelet); thromboxane A synthase 1 (platelet) CYP5 CYP5A1 FLJ52771 GHOSAL THAS TS TXAS TXS BDPLT14; thromboxane A synthase 1 (platelet, cytochrome P450, family 5, subfamily A); thromboxane synthase; Thromboxane-A synthase; TS; TXA synthase; TXS
Gene Aliases: BDPLT14; CYP5; CYP5A1; GHOSAL; TBXAS1; THAS; TS; TXAS; TXS
UniProt ID: (Bovine) Q2KIG5, (Human) P24557, (Pig) P47787, (Mouse) P36423, (Rat) P49430
Entrez Gene ID: (Dog) 482771, (Bovine) 509326, (Rabbit) 100339354, (Horse) 100066198, (Human) 6916, (Pig) 397112, (Mouse) 21391, (Guinea pig) 100719442, (Rat) 24886
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.