Invitrogen
Manufacturer Code:PA5112742
Catalog # PIPA5112742
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Mouse Thrombospondin-3 protein (24-350AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: THBS3 Thbs-3 thrombospondin 3 Thrombospondin-3 Tsp3; Thrombospondin-3
Gene Aliases: Thbs-3; THBS3; TSP3
UniProt ID: (Human) P49746, (Mouse) Q05895
Entrez Gene ID: (Human) 7059, (Mouse) 21827
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.