Novus Biologicals
Manufacturer Code:NB1002059SS
Catalog # NB1002059SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | A6.1 |
Immunogen | BALB/C mice were injected with reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Glycoprotein G; THBS THBS-1 thrombospondin 1 thrombospondin-1p180 TSP1thrombospondin-1 TSPTSP-1; Thrombospondin-1; thrombospondin-1p180; thrombospondin-p50
Gene Aliases: THBS; THBS-1; THBS1; TSP; TSP-1; TSP1
UniProt ID: (Human) P07996
Entrez Gene ID: (Human) 7057
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.