Invitrogen
Manufacturer Code:MA513377
Catalog # PIMA513377
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | A4.1 |
Immunogen | Calcium-replete native purified human TSP from the supernatant of thrombin-activated platelets |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Glycoprotein G; Thrombospondin-1; thrombospondin-1 thrombospondin-1p180; thrombospondin-1p180; thrombospondin-p50
Gene Aliases: tbsp1; THBS; THBS-1; THBS1; thrombospondin-1; TSP; TSP-1; TSP1
UniProt ID: (Human) P07996, (Bovine) Q28178
Entrez Gene ID: (Human) 7057, (Chicken) 373987, (Bovine) 281530, (Rat) 445442, (Mouse) 21825
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.