R&D Systems
Manufacturer Code:MAB78591SP
Catalog # MAB78591SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 899642R |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Thrombospondin-1 Asn19-Ser1171 Accession # P35441 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
Protein Aliases: Glycoprotein G; THBS THBS1 THBS-1 thrombospondin 1 Thrombospondin1 thrombospondin-1p180 TSP-1 TSP1thrombospondin-1 TSPTSP-1; Thrombospondin-1; thrombospondin-1p180; thrombospondin-p50
Gene Aliases: THBS; THBS-1; THBS1; TSP; TSP-1; TSP1
UniProt ID: (Human) P07996
Entrez Gene ID: (Human) 7057
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.