Invitrogen
Manufacturer Code:PA595445
Catalog # PIPA595445
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human Thrombospondin recombit protein (Position: D27-H112). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycoprotein G; Thrombospondin-1; Thrombospondin-1 THBS1 TSP TSP1; thrombospondin-1p180; thrombospondin-p50
Gene Aliases: tbsp1; THBS; THBS-1; THBS1; TSP; TSP-1; TSP1
UniProt ID: (Human) P07996
Entrez Gene ID: (Human) 7057, (Rat) 445442, (Mouse) 21825
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.