Invitrogen
Manufacturer Code:MA529627
Catalog # PIMA529627
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 54 |
Immunogen | Recombit Human Thioredoxin-2/TXN2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: mitochondrial thioredoxin; mitochondrial thioredoxin MT-TRX MTRX; MTRX; Thioredoxin, mitochondrial; Thioredoxin-2
Gene Aliases: COXPD29; MT-TRX; MTRX; TRX2; TXN2
UniProt ID: (Human) Q99757
Entrez Gene ID: (Human) 25828
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.