Novus Biologicals
Manufacturer Code:NBP29012350UL
Catalog # NB021710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 54 |
Immunogen | Recombinant Human Thioredoxin-2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: mitochondrial thioredoxin; mitochondrial thioredoxin MTRX MT-TRX thioredoxin 2 thioredoxin mitochondrial thioredoxin-2 TRX2; MTRX; Thioredoxin, mitochondrial; Thioredoxin-2
Gene Aliases: COXPD29; MT-TRX; MTRX; TRX2; TXN2
UniProt ID: (Human) Q99757
Entrez Gene ID: (Human) 25828
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.