Invitrogen
Manufacturer Code:PA517411
Catalog # PIPA517411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the sequence of mouse thioredoxin 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADF; ADF ATL-derived factor SASP surface-associated sulphydryl protein; ATL-derived factor; Thioredoxin; Trx
Gene Aliases: ADF; AW550880; Trx1; Txn; Txn1
UniProt ID: (Rat) P11232, (Mouse) P10639
Entrez Gene ID: (Rat) 116484, (Mouse) 22166
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.