R&D Systems
Manufacturer Code:MAB31201100
Catalog # MAB31201100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 353817 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human TfR2 Arg105-Phe801 Accession &num Q9UP52 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HFE3 HFE3transferrin receptor protein 2 MGC126368 TfR2 TFRC2 transferrin receptor 2; TfR2; Transferrin receptor protein 2
Gene Aliases: HFE3; TFR2; TFRC2
UniProt ID: (Human) Q9UP52
Entrez Gene ID: (Human) 7036
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.