Novus Biologicals
Manufacturer Code:NBP293436100UL
Catalog # NB028748
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 582-801 of human TFR2 (NP_003218.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HFE3transferrin receptor protein 2 MGC126368 TfR2 TFRC2 transferrin receptor 2; TfR2; Transferrin receptor protein 2
Gene Aliases: HFE3; TFR2; TFRC2
UniProt ID: (Human) Q9UP52
Entrez Gene ID: (Human) 7036
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.