Novus Biologicals
Manufacturer Code:NBP289742100UL
Catalog # NB020965
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | Recombinant Human TfR (Transferrin R) protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CD71; CD71 antigen CD71TRFR p90 T9 TfR TFR1 TR transferrin receptor (p90 CD71) transferrin receptor protein 1 Trfr; p90; sTfR; T9; TR; Transferrin receptor protein 1; Transferrin receptor protein 1, serum form
Gene Aliases: CD71; IMD46; p90; T9; TFR; TFR1; TFRC; TR; TRFR
UniProt ID: (Human) P02786
Entrez Gene ID: (Human) 7037
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.