Novus Biologicals
Manufacturer Code:NBP184129
Catalog # NBP184129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:EEPLNLEGLVAHRVAGVQAREKQPAEPPAPLRRRAASDGQYENQSPEATSPRSPGVRSPVQCVSPELALTIALNPGGRPKEPHLHSYK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp586K0617 matrix-remodelling associated 6 Matrix-remodelling-associated protein 6 MGC88584 MST091 MST122 MST127 MSTP127 MXRA6 tensin tensin 1 tensin-1 TNSMSTP122; matrix-remodelling associated 6; Matrix-remodelling-associated protein 6; protein phosphatase 1, regulatory subunit 155; Tensin-1
Gene Aliases: MST091; MST122; MST127; MSTP091; MSTP122; MSTP127; MXRA6; PPP1R155; TNS; TNS1
UniProt ID: (Human) Q9HBL0
Entrez Gene ID: (Human) 7145
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.