R&D Systems
Manufacturer Code:AF4578SP
Catalog # AF4578SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Teneurin-2 Met1-Lys183 Accession # AAI72353 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: neurestin alpha; odd Oz/ten-m homolog 2; odz, odd Oz/ten-m homolog 2; Protein Odd Oz/ten-m homolog 2; Ten-2; Ten-2 ICD; Ten-2 intracellular domain; Ten-2, soluble form; Ten-m2; Tenascin-M2; Teneurin transmembrane protein 2; Teneurin-2; teneurin-2 ODZ2 Ten-2 Tenascin-M2 teneurin transmembrane protein 2 Teneurin2 Ten-m2 TNM2
Gene Aliases: KIAA1127; ODZ2; ten-2; TEN-M2; TENM2; TNM2
UniProt ID: (Human) Q9NT68
Entrez Gene ID: (Human) 57451
Molecular Function:
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.