Novus Biologicals
Manufacturer Code:NBP267479
Catalog # NBP267479
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SU36-01 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: 150-225 Cytotactin Glioma-associated-extracellular matrix antigen GMEM GP 150-225 Hexabrachion hexabrachion (tenascin C cytotactin) hexabrachion (tenascin) HXBcytotactin JI MGC167029 Myotendinous antigen neuronectin tenascin tenascin C Tenascin-C tenascin-C isoform 14/AD1/16 TN-C TNGP; Cytotactin; deafness, autosomal dominant 56; Glioma-associated-extracellular matrix antigen; GMEM; GP 150-225; Hexabrachion; hexabrachion (tenascin); JI; Myotendinous antigen; Neuronectin; Tenascin; Tenascin-C; tenascin-C additional domain 1; tenascin-C isoform 14/AD1/16; TN; TN-C
Gene Aliases: 150-225; DFNA56; GMEM; GP; HXB; JI; TN; TN-C; TNC
UniProt ID: (Human) P24821
Entrez Gene ID: (Human) 3371
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.