Invitrogen
Manufacturer Code:PA580172
Catalog # PIPA580172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of mouse TNR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Janusin; Neural recognition molecule J1-160/180; Restrictin; tenascin R (restrictin, janusin); Tenascin-R; Tenascin-R (Restrictin janusin J1-160/180); Tenascin-R (Restrictin, janusin, J1-160/180); Tenascin-R TN-R Janusin Neural recognition molecule J1-160/180 Restrictin Tnr; TN-R
Gene Aliases: J1-tenascin; J1NRM; janusin; restrictin; TN-R; TNR
UniProt ID: (Human) Q92752, (Mouse) Q8BYI9, (Rat) Q05546
Entrez Gene ID: (Human) 7143, (Mouse) 21960, (Rat) 25567
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.