Invitrogen
Manufacturer Code:PA531143
Catalog # PIPA531143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 762 and 1023 of Human TNR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Janusin; Neural recognition molecule J1-160/180; Restrictin; Tenascin-R; TN-R; TN-R janusin restrictin tenascin-R
Gene Aliases: J1-tenascin; janusin; restrictin; TN-R; Tnr
UniProt ID: (Mouse) Q8BYI9
Entrez Gene ID: (Mouse) 21960
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.