Invitrogen
Manufacturer Code:MA516086
Catalog # PIMA516086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4C8MS |
Immunogen | Recombit human Tenascin C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 150-225 GMEM GP HXB JI TN TN-C GP 150-225 cytotactin glioma-associated-extracellular matrix antigen hexabrachion (tenascin) myotendinous antigen neuronectin tenascin tenascin-C isoform 14/AD1/16; Cytotactin; deafness, autosomal dominant 56; Glioma-associated-extracellular matrix antigen; GMEM; GP 150-225; Hexabrachion; hexabrachion (tenascin); JI; Myotendinous antigen; Neuronectin; Tenascin; Tenascin-C; tenascin-C additional domain 1; tenascin-C isoform 14/AD1/16; TN; TN-C; tnc
Gene Aliases: 150-225; AI528729; C130033P17Rik; cytotactin; DFNA56; GMEM; GP; HXB; JI; Ten; tenascin-C; TN; TN-C; TNC
UniProt ID: (Human) P24821, (Mouse) Q80YX1
Entrez Gene ID: (Human) 3371, (Rat) 116640, (Mouse) 21923
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.