Invitrogen
Manufacturer Code:MA511466
Catalog # PIMA511466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SEN28 |
Immunogen | Recombit protein corresponding to the amino terminal region of the TdT molecule |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: deoxynucleotidyltransferase, terminal; DNA nucleotidylexotransferase; DNA nucleotidylexotransferase nucleosidetriphosphate:DNA deoxynucleotidylexotransferase terminal addition enzyme terminal deoxynucleotidyltransferase terminal deoxyribonucleotidyltransferase terminal transferase; nucleosidetriphosphate:DNA deoxynucleotidylexotransferase; Terminal addition enzyme; Terminal deoxynucleotidyltransferase; terminal deoxyribonucleotidyltransferase; Terminal transferase
Gene Aliases: DNTT; TDT
UniProt ID: (Human) P04053
Entrez Gene ID: (Human) 1791
Molecular Function:
DNA binding protein
DNA-directed DNA polymerase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.