Invitrogen
Manufacturer Code:14973980
Catalog # 14-973-980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7UNA8U6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: deoxynucleotidyltransferase terminal DNA nucleotidylexotransferase DNTT nucleosidetriphosphate:DNA deoxynucleotidylexotransferase Tdt terminal addition enzyme terminal deoxynucleotidyl transferase terminal deoxynucleotidyltransferase terminal deoxyribonucleotidyltransferase terminal transferase; deoxynucleotidyltransferase, terminal; DNA nucleotidylexotransferase; nucleosidetriphosphate:DNA deoxynucleotidylexotransferase; TDT; Terminal addition enzyme; terminal deoxynucleotidyl transferase; Terminal deoxynucleotidyltransferase; terminal deoxyribonucleotidyltransferase; Terminal transferase
Gene Aliases: DNTT; TDT
UniProt ID: (Human) P04053, (Mouse) P09838
Entrez Gene ID: (Human) 1791, (Mouse) 21673, (Rat) 294051
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.