Abnova
Manufacturer Code:MAB18722
Catalog # 89-338-647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 23c |
Immunogen | Purified recombinant mouse TCP1 alpha construct encoding the C-terminal half of the 1.8 kb full-length Tcp 1b gene expressed in E. coli. Detects the COOH group. |
Conjugate | ATTO700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: AI528772; c-cpn; CCT; CCT-alpha; Cct1; Ccta; p63; t-complex polypeptide 1; T-complex protein 1 subunit alpha; T-complex protein 1 subunit alpha A; T-complex protein 1 subunit alpha B; Tailless complex polypeptide 1A; Tailless complex polypeptide 1B; Tcp-1; TCP-1-A; TCP-1-alpha; TCP-1-B; Tp63; TRic
Gene Aliases: AI528772; c-cpn; CCT; Cct1; Ccta; p63; Tcp-1; Tcp1; Tp63; TRic
UniProt ID: (Mouse) P11983
Entrez Gene ID: (Mouse) 21454
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.