Novus Biologicals
Manufacturer Code:NBP24240225UG
Catalog # NBP24240225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9G3 |
Immunogen | aa 12-24 (pTyr18) for Tau protein was used as the immunogen for this antibody. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Lyophilized antibody can be kept at 4°C for up to 3 months and should be kept at -20°C for long-term storage. To avoid freeze-thaw cycles reconstituted antibody should be aliquoted before freezing for short-term storage (-20°C) or for lon |
Mouse Monoclonal Antibody
Protein Aliases: DDPAC FLJ31424 FTDP-17 G protein beta1/gamma2 subunit-interacting factor 1 MGC138549 microtubule-associated protein tau MSTDMAPTL MTBT1Neurofibrillary tangle protein MTBT2 PHF-tau PPND tau TAUPaired helical filament-tau; G protein beta1/gamma2 subunit-interacting factor 1; Microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; Neurofibrillary tangle protein; Paired helical filament-tau; PHF-tau; protein phosphatase 1, regulatory subunit 103
Gene Aliases: DDPAC; FTDP-17; MAPT; MAPTL; MSTD; MTBT1; MTBT2; PPND; PPP1R103; TAU
UniProt ID: (Human) P10636
Entrez Gene ID: (Human) 4137
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.