Novus Biologicals
Manufacturer Code:NB10082247
Catalog # NB10082247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The antiserum was produced against synthesized non-phosphopeptide derived from human Tau around aa. 231 (V-R-T-P-P) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DDPAC FLJ31424 FTDP-17 G protein beta1/gamma2 subunit-interacting factor 1 MGC138549 microtubule-associated protein tau MSTDMAPTL MTBT1Neurofibrillary tangle protein MTBT2 PHF-tau PPND tau TAUPaired helical filament-tau; G protein beta1/gamma2 subunit-interacting factor 1; Microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; Neurofibrillary tangle protein; Paired helical filament-tau; PHF-tau; protein phosphatase 1, regulatory subunit 103
Gene Aliases: DDPAC; FTDP-17; MAPT; MAPTL; MSTD; MTBT1; MTBT2; PPND; PPP1R103; TAU
UniProt ID: (Human) P10636
Entrez Gene ID: (Human) 4137
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.