Invitrogen
Manufacturer Code:PA568624
Catalog # PIPA568624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human Tankyrase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 5; ARTD5; PARP-5a PARP5A PARPL TIN1 TINF1 TNKS1 pART5 TANK1 TNKS-1 TRF1-interacting ankyrin-related ADP-ribose polymerase poly &lsqbADP-ribose] polymerase 5A tankyrase I tankyrase-1; Poly [ADP-ribose] polymerase 5A; Poly [ADP-ribose] polymerase tankyrase-1; Protein poly-ADP-ribosyltransferase tankyrase-1; TANK1; tankyrase 1; Tankyrase I; tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase; Tankyrase-1; TNKS-1; TRF1-interacting ankyrin-related ADP-ribose polymerase; TRF1-interacting ankyrin-related ADP-ribose polymerase 1
Gene Aliases: 4930554K12Rik; AI662855; ARTD5; C86528; D130072O21Rik; mTNKS1; PARP-5a; PARP5A; PARPL; pART5; TANK1; TIN1; TINF1; TNKS; TNKS1
UniProt ID: (Human) O95271, (Mouse) Q6PFX9
Entrez Gene ID: (Horse) 100058499, (Bovine) 535030, (Dog) 475603, (Rabbit) 100351292, (Human) 8658, (Mouse) 21951, (Guinea pig) 100728577, (Rat) 290794
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.