Invitrogen
Manufacturer Code:PA5111075
Catalog # PIPA5111075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human Tankyrase 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 6; ADP-ribosyltransferase diphtheria toxin-like 6 ARTD6 PARP5B PARP-5b PARP5C PARP-5c pART6 Poly [ADP-ribose] polymerase 5B Poly [ADP-ribose] polymerase tankyrase-2 Protein poly-ADP-ribosyltransferase tankyrase-2 TANK2 tankyrase 2 Tankyrase II Tankyrase-2 Tankyrase-like protein Tankyrase-related protein TNKL TNKS2 TNKS-2 TRF1-interacting ankyrin-related ADP-ribose polymerase 2; ARTD6; Poly [ADP-ribose] polymerase 5B; Poly [ADP-ribose] polymerase tankyrase-2; Protein poly-ADP-ribosyltransferase tankyrase-2; TANK2; Tankyrase II; tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase 2; Tankyrase-2; Tankyrase-like protein; Tankyrase-related protein; TNKS-2; TRF1-interacting ankyrin-related ADP-ribose polymerase 2
Gene Aliases: ARTD6; PARP-5b; PARP-5c; PARP5B; PARP5C; pART6; TANK2; TNKL; TNKS2
UniProt ID: (Human) Q9H2K2
Entrez Gene ID: (Human) 80351
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.