Novus Biologicals
Manufacturer Code:NB100186AF488
Catalog # NB100186X
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to a region of human Tankyrase within residues 250-350 [UniProt# O95271]. The actual peptide sequence is proprietary. |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 5; ARTD5; EC 2.4.2.30 PARP-5a PARP5APARPL pART5 Poly [ADP-ribose] polymerase 5A Tankyrase I tankyrase TRF1-interacting ankyrin-related ADP-ribose polymerase tankyrase-1 TIN1TANK1 TINF1TNKS-1 TNKS1TRF1-interacting ankyrin-related ADP-ribose polymerase; Poly [ADP-ribose] polymerase 5A; Poly [ADP-ribose] polymerase tankyrase-1; Protein poly-ADP-ribosyltransferase tankyrase-1; TANK1; Tankyrase I; tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase; Tankyrase-1; TNKS-1; TRF1-interacting ankyrin-related ADP-ribose polymerase; TRF1-interacting ankyrin-related ADP-ribose polymerase 1
Gene Aliases: ARTD5; PARP-5a; PARP5A; PARPL; pART5; TIN1; TINF1; TNKS; TNKS1
UniProt ID: (Human) O95271
Entrez Gene ID: (Human) 8658
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.