Novus Biologicals
Manufacturer Code:NBP250319AF700
Catalog # NBP250319AI
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 93000000000000 |
Immunogen | Recombinant protein fragment |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ILWEQ KIAA1027 talin 1 talin-1 TLN; Talin-1
Gene Aliases: ILWEQ; KIAA1027; TLN; TLN1
UniProt ID: (Human) Q9Y490
Entrez Gene ID: (Human) 7094
Molecular Function: actin family cytoskeletal protein cell adhesion molecule cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.