Novus Biologicals
Manufacturer Code:NBP298750AF750
Catalog # NB105490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Tafazzin/TAZ. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTHSEFE cardiomyopathy dilated 3A (X-linked) CMD3A EFE2FLJ27390 G4.5endocardial fibroelastosis 2 LVNCX Protein G4.5 tafazzin Taz1 XAP-2; Protein G4.5; Tafazzin
Gene Aliases: BTHS; CMD3A; EFE; EFE2; G4.5; LVNCX; TAZ; Taz1
UniProt ID: (Human) Q16635
Entrez Gene ID: (Human) 6901
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.