Invitrogen
Manufacturer Code:PA543859
Catalog # PIPA543859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TYW3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: tRNA wybutosine-synthesizing protein 3 homolog; tRNA(Phe) 7-((3-amino-3-carboxypropyl)-4-demethylwyosine(37)-N(4))-methyltransferase; tRNA-yW synthesizing protein 3 homolog (S. cerevisiae); tRNA-yW-synthesizing protein 3
Gene Aliases: 5230400J09Rik; C1orf171; RGD1559623; TYW3
UniProt ID: (Human) Q6IPR3, (Bovine) Q5E9U4, (Mouse) Q8BSA9
Entrez Gene ID: (Horse) 100053370, (Rabbit) 100348497, (Human) 127253, (Bovine) 519731, (Rat) 499731, (Mouse) 209584
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.