Novus Biologicals
Manufacturer Code:NBP176584
Catalog # NBP176584
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 17 amino acid synthetic peptide near the center of human TYW3. The immunogen is located within amino acids 160 - 210 of TYW3. Amino Acid Squence: ANQKMEENKKRIERFYN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf171 chromosome 1 open reading frame 171 EC 2.1.1 EC 2.1.1.- FLJ40918 tRNA wybutosine-synthesizing protein 3 homolog tRNA-yW synthesizing protein 3 homolog (S. cerevisiae) tRNA-yW-synthesizing protein 3; tRNA wybutosine-synthesizing protein 3 homolog; tRNA(Phe) 7-((3-amino-3-carboxypropyl)-4-demethylwyosine(37)-N(4))-methyltransferase; tRNA-yW-synthesizing protein 3
Gene Aliases: C1orf171; TYW3
UniProt ID: (Human) Q6IPR3
Entrez Gene ID: (Human) 127253
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.