Invitrogen
Manufacturer Code:PA556435
Catalog # PIPA556435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TYSND1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NET41 peroxisomal leader peptide-processing protease trypsin domain-containing protein 1; peroxisomal cysteine endopeptidase; Peroxisomal leader peptide-processing protease; Peroxisomal leader peptide-processing protease, 15 kDa form; Peroxisomal leader peptide-processing protease, 45 kDa form; Trypsin domain-containing protein 1
Gene Aliases: NET41; TYSND1
UniProt ID: (Human) Q2T9J0
Entrez Gene ID: (Human) 219743, (Rat) 365571
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.